The Genova Nano can measure small sample volumes, conserves precious samples, reduces the need for dilutions and eliminates the requirement for cuvettes. Cleaning is quick and simple; wiping the read head with a microfibre cloth removes all trace of the sample, allowing faster change over between samples and therefore increasing sample throughput.
- Icon driven software for intuitive navigation with method and result saving to USB stick
- Small footprint with graphical display in the compartment lid (optional internal printer)
- Pre-programmed parameters for DNA/RNA analysis; methods for protein analysis ; purity scan function ; plus standard spectrophotometer functions
The Genova Nano functions as a dedicated life science analyser, allowing the measurement of DNA concentrations and purity ratios using wavelengths recorded at 260, 280 and 230 nm with an optional correction at 320 nm. It is pre-programmed with Pierce 660, Bradford, Lowry, Biuret, BCA and direct UV methods for protein analysis. The Genova Nano has an OD measurement mode enabling cell growth to be monitored by measuring absorbance at 600 nm, 595 nm or any other user selected wavelength. The purity scan across the entire wavelength range from 198 to 1000 nm (interval: selectable 1, 2 or 5 nm) displays any distorted peaks enabling impurities to be easily identified.
The instrument can also be used as a standard spectrophotometer with measurement modes for photometrics, concentration, multi-wavelength, spectrum scanning, quantitation and kinetics.
Informace o dodávce: Supplied fitted with micro-volume accessory (quartz stainless steel), 10×10 mm cuvette holder and four GB USB stick.