Tisk…
Digital thermometer
Dodavatel: Hecht
HECH43232010EA 1470 CZK
HECH43232010
Digital thermometer
Teploměry Teploměry digitální
Versatile thermometer with stainless steel probe and fixed extension cable.
  • LCD display
  • Maximum/minimum display with temperature memory
  • Hi-Lo alarm, programmable
  • Low battery indicator
Informace o dodávce: Battery (1,5 V AAA) and instructions for use.