Tisk…
LABOCAP caps, for test tubes
Dodavatel: Hecht
HECH42777120 HECH42777104 HECH42777116
LABOCAP caps, for test tubes
Aluminium
  • Made of anodised aluminium with inner stainless steel spring
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
With handle. Cap for sterile but not hermetic closing of test tubes and other vessels.