Tisk…

(S)-(-)-3-Aminoquinuclidine dihydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL415723-1GEA 4080 CZK
SIAL415723-1G
(S)-(-)-3-Aminoquinuclidine dihydrochloride, Sigma-Aldrich®
(S)-(-)-3-Aminoquinuclidine dihydrochloride
Vzorec: C₇H₁₆Cl₂N₂
MW: 199,1213 g/mol
MDL Number: MFCD00191753
CAS číslo: 119904-90-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR