Tisk…

2-Fluoro-5-(pentafluorothio)benzaldehyde ≥97%

Varování

H33481.MDEA 1910 CZK
H33481.MD H33481.03
2-Fluoro-5-(pentafluorothio)benzaldehyde ≥97%
2-Fluoro-5-(pentafluorothio)benzaldehyde
Vzorec: C₇H₄F₆OS
MW: 250,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652493
CAS číslo: 1240257-02-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR