Tisk…

Acetyl anthranil, Sigma-Aldrich®

Dodavatel: Merck

SIAL457752-10GEA 2460 CZK
SIAL457752-10G
Acetyl anthranil, Sigma-Aldrich®
Acetyl anthranil
Vzorec: C₉H₇NO₂
CAS číslo: 525-76-8
EINECS: 208-381-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR