Blue Line autosampler syringes for 7693A ALS, with fitted plungers
Supplier: Agilent
Certificates
About this item
These autosampler syringes are used for homogenous liquid samples.
- Plungers are not interchangeable or replaceable
Each fitted syringe is individually matched with the plunger for precision injection.
Plungers are suitable for gas and liquid samples, the PTFE tip of the plunger creates a tight seal between the plunger and glass, helping to reduce carry-over and increase syringe lifetime.
Ordering information: Replacement plungers available.
Product Information
- Catalog #Supplier #GaugeVolumeLengthType of tipTypePkAvailabilityPrice
- HEWLG4513-80212G4513-8021226s5 µl42 mmHPStraight, fixed needle6Log in for availability1 * 6 KS22 800,00 KčSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG4513-80206G4513-8020623-26s5 µl42 mmHPTapered, fixed needle1Log in for availability1 * 1 KS4 350,00 KčSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG4513-80209G4513-802092310 µl42 mmHPStraight, fixed needle1Log in for availability1 * 1 KS2 600,00 KčSpecifications:
Accessories
Specifications
Environmentally Preferable - HEWLG4513-80204G4513-8020423-26s10 µl42 mmHPTapered, fixed needle1Log in for availability1 * 1 KS2 800,00 KčSpecifications: