Tisk…
Bunsen burner, Labogaz®
Dodavatel: Hecht
HECH41396010EA 2500 CZK
HECH41396010
Bunsen burner, Labogaz®
Ohřívače Hořáky plynové
Bunsen burner with new assembling-system for more security to cartridges without safety valve, gas consumption 55 g/h.