Tisk…
Stand magnifier
Dodavatel: Eschenbach
ESCH2626EA 1360 CZK
ESCH2626
Stand magnifier
Aspheric magnifier with a modern white plastic frame and an easy access to view material.
  • PXM® lightweight lens
  • Ceratec® scratch resistant coating
  • Open transparent plastic base for writing