Centrifugal filters, Centricon® Plus-70
New Product
Centrifugal filter units concentrate samples in the range of 15 ml to 70 ml. The large dead stop volume of 350 µl prevents spinning samples to dryness. It has two vertical regenerated cellulose membranes for fast, efficient tangential flow.
- Available with different pore size membranes
- 90% typical recovery and low hold-up volume
- PP housing minimises non-specific protein binding
Applications includes concentration and desalting of chromatography column eluates, concentrating monoclonal antibodies, concentrating proteins or viruses from culture supernatants, clarifying tissue homogenates and lysates, buffer exchange or diafiltration.
- Catalog No:
- MILYUFC710008
- MILYUFC703008
- MILYUFC700308
- MILYUFC701008
- Ø×H:
- 60×121 mm
- 60×121 mm
- 60×121 mm
- 60×121 mm
- Sample volume:
- 15000 - 70000 µl
- 15000 - 70000 µl
- 15000 - 70000 µl
- 15000 - 70000 µl
- MWCOMembraneTypeColour codeFiltration area
- MILYUFC700308UFC7003083 kDRCUltrafiltrationClear19 cm²1 * 8 KS8 600,00 Kč: UFC700308: MILYUFC700308New Product
Specifications
Environmentally PreferableØ×H60×121 mmSample volume15000 - 70000 µlNew Product - MILYUFC701008UFC70100810 kDRCUltrafiltrationClear19 cm²1 * 1 KS8 760,00 Kč: UFC701008: MILYUFC701008New Product
Specifications
Environmentally PreferableØ×H60×121 mmSample volume15000 - 70000 µlNew Product - MILYUFC703008UFC70300830 kDRCUltrafiltrationClear19 cm²1 * 8 KS8 760,00 Kč: UFC703008: MILYUFC703008New Product
Specifications
Environmentally PreferableØ×H60×121 mmSample volume15000 - 70000 µlNew Product - MILYUFC710008UFC710008100 kDRCUltrafiltrationClear19 cm²1 * 8 KS8 760,00 Kč: UFC710008: MILYUFC710008New Product
Specifications
Environmentally PreferableØ×H60×121 mmSample volume15000 - 70000 µlNew Product