Tisk…
Alcohol hand gels, Spirigel®
Dodavatel: Ecolab
Spirigel®
SHIE3030550EA 0 CZK
SHIE3030550
Alcohol hand gels, Spirigel®
Péče o pokožku Dezinfekce na ruce Gelové dezinfekční prostředky na pleť
70% denatured ethanol gel for the rapid disinfection of physically clean hands.
  • Proven efficacy against gram positive and gram negative bacteria including E.coli and S.epidermidis; fast acting with proven efficacy against EN 1500 in 30 seconds and removes 99,9 % of transient organisms
  • Hand gel evaporates quickly and leaves no sticky residue
  • new Spiripump has been developed to eliminate clogging and ensure a controlled delivery of the alcohol gel without splashing
  • Pump dispenser delivers the correct measure for each hand sanitisation procedure and avoids wastage
Certifikace: EN 12054, EN 1500