Tisk…
Microcentrifuge, refrigerated, 5427 R (IVD marked)
Dodavatel: Eppendorf
521-1763EA 181920 CZK
521-1763 521-1730 521-1729 521-1888 521-1731 521-1727 521-1728 521-1726
Microcentrifuge, refrigerated, 5427 R (IVD marked)
Centrifugy Mikrocentrifugy
This fast running microcentrifuge is designed for maximum application flexibility and offers a versatile choice of rotors in order to serve different user demands.
  • Short-Spin function allows for quick short centrifugation with pre-selectable rotation speed
  • Very quiet operation due to further improved sound insulation
  • FastTemp for rapid pre-cooling of the centrifuge, e.g. from 23 to 4 °C in approx. 11 minutes
  • Continuous cooling keeps the temperature after the run constant
  • Alternatively to continuous cooling: ECO-shut off of the compressor (adjustable after 1, 2, 4 or 8 hours of non use) for a lower energy consumption
The available rotors include high capacity rotors with up to 48 places as well as high speed rotors for a quick sample throughput.
Informace pro objednávání: Can be supplied with or without rotor.