Tisk…
HPLC columns, Pursuit™
Dodavatel: Agilent Technologies
Pursuit™
VARIA3001050X030EA 19340 CZK
VARIA3001050X030 VARIA3001100C030 VARIA3000100C030 VARIA3041030X020 VARIA3040030X020 VARIA3040100X212 VARIA3051150X030 VARIA3050150X030 VARIA3050200X046 VARIA3032250X046 VARIA3040250X100 VARIA3030150C046 VARIA3031150C046 VARIA3041050X030 VARIA7000150X046 VARIA3041200X020 VARIA3031CG3 VARIA3031CG2 VARIA3000100C020 VARIA3002250X212 VARIA3031100X030 VARIA3000250X212 VARIA3000125X040 VARIA3001020X020 VARIA3031100X046 VARIA3001050X020 VARIA3001050C020 VARIA3000250C046 VARIA3001100C020 VARIA3051150X020 VARIA3000050X046 VARIA3000100X046 VARIA3050150X046 VARIA3030250X046 VARIA3030250X040 VARIA3051050X046 VARIA3041150X010 VARIA7001100X020 VARIA3040050X046 VARIA3030050X020 VARIA3002250X100 VARIA3000250X100 VARIA3001250X046 VARIA3040250C046 VARIA3002250X500 VARIA3041050X020 VARIA3000150X100 VARIA3001CG3 VARIA3001CG2 VARIA3000100X030 VARIA3001100X030 VARIA3050250X020 VARIA3040250X046 VARIA3041250X046 VARIA3041150X100 VARIA3032150X046 VARIA3000250X030 VARIA3041100X030 VARIA3051030X020 VARIA3000150X212 VARIA3041100X046 VARIA3040250C030 VARIA3040250X030 VARIA3000100X020 VARIA3050150X020 VARIA3041100P046 VARIA3050150X021 VARIA3001100X046 VARIA3051150X046 VARIA3000250X040 VARIA3031050X020 VARIA3051100X020 VARIA3030250C046 VARIA3040CG3 VARIA3040CG2 VARIA3000250X046 VARIA3040100X020 VARIA3030100X046 VARIA3000250C030 VARIA3031020X020 VARIA3000150C046 VARIA3000150X046 VARIA3001150X046 VARIA3002300X039 VARIA3002150X046 VARIA3041250X020 VARIA3030030X046 VARIA3041150X046 VARIA3031150X020 VARIA3040150X046 VARIA3051100X030 VARIA3001150C030 VARIA3000150C030 VARIA3030150X020 VARIA3000CG2 VARIA3000150X039 VARIA3000CG3 VARIA3050100X020 VARIA3001100X020 VARIA3051100X046 VARIA7001100C046 VARIA3050250X030 VARIA3000150X030 VARIA3050050X020 VARIA3001030X046 VARIA3000250X020 VARIA3000250X021 VARIA3041100X020 VARIA7000250C046 VARIA3001150C046 VARIA3001050C030 VARIA3002150X500 VARIA3050250X100 VARIA3032250X100 VARIA3000150X020 VARIA7000250X046 VARIA3001150X020 VARIA7000150C046 VARIA3041150X020 VARIA3031150X046 VARIA7001150C046 VARIA3040150X020 VARIA3030150X046 VARIA7001100X030 VARIA3000100C046 VARIA3040050X010 VARIA3000050X020 VARIA3040100C046 VARIA7001100X046 VARIA3002150X212 VARIA3001050C046 VARIA3050100X046 VARIA3000020X020 VARIA3001050X046 VARIA3050150X100 VARIA3001150X030 VARIA3051020X020 VARIA3051050X020 VARIA3030CG2 VARIA3041150X030 VARIA3030CG3 VARIA3001250X020 VARIA3041050X046 VARIA3002250X046 VARIA3040050X020 VARIA3001030X020 VARIA3031100X020 VARIA3040050X021
HPLC columns, Pursuit™
Kolony chromatografické
These Pursuit™ columns are ideal for analysing lead compounds and biological samples. The need for ion pairing agents such as TFA is often eliminated, thus maximising the performance of single and parallel multi-channel LC/MS systems.
  • Built on the larger 200 Å pore size silica
  • For LC/MS and high throughput applications
High ligand density delivers up to 40% faster separations without sacrificing resolution by optimising mass transfer with the larger pore size.