Tisk…
Wireless digital cameras, Moticam® x and x3
Dodavatel: Motic
Moticam®
630-1840EA 10700 CZK
MOTI1100600100691 MOTI1100600101031
Wireless digital cameras, Moticam® x and x3
Kamery Kamery digitální
High resolution streaming wireless digital cameras.
  • Support six devices at the same time
  • x3 can be connected to Ethernet via an RJ 45 port
  • Live images on any computer using the Images Plus 2.0 software
View, capture or edit live images via MotiConnect App, on your smartphone or tablet (measurements, annotations) or laptop (Windows XP or higher) via the standard Images Plus software.
Informace o dodávce: All-in-one box: Two variable diameter eyepiece adapters, calibration slide, macro-tube, USB power supply and software.